CAS RN 5190-68-1
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
2
of
2
results
1
Promotions Available
4-Chloroquinazoline, 95%, Thermo Scientific™
4-Chloroquinazoline, 95%, Thermo Scientific™
2
Sigma Aldrich 4-Chloroquinazoline
MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.