accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 519056-61-2

IUPAC Name: (4-tert-butylphenyl)methanesulfonyl chloride
Synonyms: (4-tert-butylphenyl)methanesulfonyl chloride
Molecular Weight (g/mol): 246.75
Molecular Formula: C11H15ClO2S
InChi Key: NEJDUPGGBSPJLG-UHFFFAOYSA-N
SMILES: CC(C)(C)C1=CC=C(CS(Cl)(=O)=O)C=C1
Molecular Weight (g/mol) 
  • (1)
  • (1)
Quantity 
  • (2)
Grade 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results