accessibility menu, dialog, popup

UserName

Viewing profile as user:
FFFNHNNNHNOOHNN

CAS RN 519178-28-0

IUPAC Name: N6-[(4-nitrophenyl)methyl]-N2-{[3-(trifluoromethyl)phenyl]methyl}-7H-purine-2,6-diamine
Synonyms: N6-[(4-nitrophenyl)methyl]-N2-{[3-(trifluoromethyl)phenyl]methyl}-7H-purine-2,6-diamine
Molecular Weight (g/mol): 443.39
Molecular Formula: C20H16F3N7O2
InChi Key: DDSBPUYZPWNNGH-UHFFFAOYSA-N
SMILES: [O-][N+](=O)C1=CC=C(CNC2=C3NC=NC3=NC(NCC3=CC(=CC=C3)C(F)(F)F)=N2)C=C1
Quantity 
  • (1)
  • (1)
Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results