CAS RN 5192-04-1
IUPAC Name:
1H-indol-7-amine
Synonyms:
1H-indol-7-amine
Molecular Weight (g/mol):
132.17
Molecular Formula:
C8H8N2
InChi Key:
WTFWZOSMUGZKNZ-UHFFFAOYSA-N
SMILES:
NC1=C2NC=CC2=CC=C1
1
–
1
of
1
results
1
Promotions Available
7-Aminoindole, 95%, Thermo Scientific™
7-Aminoindole, 95%, Thermo Scientific™
CAS: 5192-04-1 Molecular Formula: C8H8N2 Molecular Weight (g/mol): 132.16 InChI Key: WTFWZOSMUGZKNZ-UHFFFAOYSA-N Synonym: 2-chloro-6-hydrazinopyridine,2-chloro-6-hydrazinylpyridine,6-chloro-2-pyridyl hydrazine,6-chloro-2-pyridylhydrazine,2 1h-pyridinone, 6-chloro-, hydrazone,pubchem12983,acmc-1awi0,2-hydrazino-6-chloropyridine,6-chloro-2-hydrazinopyridine,6-chloro-2-hydrazonopyridine PubChem CID: 320004 IUPAC Name: 1H-indol-7-amine SMILES: NC1=C2NC=CC2=CC=C1