accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 5192-62-1

IUPAC Name: 2-hydroxy-3,6-dimethyl-4H-pyran-4-one
Synonyms: 2-hydroxy-3,6-dimethylpyran-4-one
Molecular Weight (g/mol): 140.14
Molecular Formula: C7H8O3
InChi Key: RJXDKSBBQBZBQJ-UHFFFAOYSA-N
SMILES: CC1=CC(=O)C(C)=C(O)O1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results