CAS RN 51963-61-2
Adenosine-5'-Triphosphate Disodium Salt Hydrate MP Biomedicals
CAS: 51963-61-2 Molecular Formula: C10H20N5Na2O16P3 Molecular Weight (g/mol): 605.19 MDL Number: MFCD00150754 InChI Key: MWEQTWJABOLLOS-MVHBFMHHNA-L PubChem CID: 134129625 SMILES: O.O.O.[Na+].[Na+].NC1=C2N=CN([C@@H]3O[C@H](COP(O)(=O)OP([O-])(=O)OP(O)([O-])=O)[C@@H](O)[C@H]3O)C2=NC=N1
Adenosine-5'-Triphosphate Disodium Salt Hydrate MP Biomedicals
CAS: 51963-61-2 Molecular Formula: C10H20N5Na2O16P3 Molecular Weight (g/mol): 605.19 MDL Number: MFCD00150754 InChI Key: MWEQTWJABOLLOS-MVHBFMHHNA-L Synonym: adenosine 5'-triphosphate disodium salt trihydrate,adenosine 5'-triphosphate disodium salt,adenosine 5'-triphosphate disodium salt trihydrate, disodium salt PubChem CID: 134129625 IUPAC Name: disodium trihydrate hydrogen [({[(2R,3S,4R,5R)-5-(6-amino-9H-purin-9-yl)-3,4-dihydroxyoxolan-2-yl]methoxy}(hydroxy)phosphoryl phosphonato)oxy]phosphonate SMILES: O.O.O.[Na+].[Na+].NC1=C2N=CN([C@@H]3O[C@H](COP(O)(=O)OP([O-])(=O)OP(O)([O-])=O)[C@@H](O)[C@H]3O)C2=NC=N1
Adenosine-5'-Triphosphate Hydrate, MP Biomedicals™
CAS: 51963-61-2 Molecular Formula: C10H20N5Na2O16P3 Molecular Weight (g/mol): 605.19 MDL Number: MFCD00150754 InChI Key: MWEQTWJABOLLOS-MVHBFMHHNA-L Synonym: adenosine 5'-triphosphate disodium salt trihydrate,adenosine 5'-triphosphate disodium salt,adenosine 5'-triphosphate disodium salt trihydrate, disodium salt PubChem CID: 134129625 IUPAC Name: disodium trihydrate hydrogen [({[(2R,3S,4R,5R)-5-(6-amino-9H-purin-9-yl)-3,4-dihydroxyoxolan-2-yl]methoxy}(hydroxy)phosphoryl phosphonato)oxy]phosphonate SMILES: O.O.O.[Na+].[Na+].NC1=C2N=CN([C@@H]3O[C@H](COP(O)(=O)OP([O-])(=O)OP(O)([O-])=O)[C@@H](O)[C@H]3O)C2=NC=N1
Adenosine-5'-Triphosphate Trihydrate, MP Biomedicals™
CAS: 51963-61-2 Molecular Formula: C10H20N5Na2O16P3 Molecular Weight (g/mol): 605.19 MDL Number: MFCD00150754 InChI Key: MWEQTWJABOLLOS-MVHBFMHHNA-L Synonym: adenosine 5'-triphosphate disodium salt trihydrate,adenosine 5'-triphosphate disodium salt,adenosine 5'-triphosphate disodium salt trihydrate, disodium salt PubChem CID: 134129625 IUPAC Name: disodium trihydrate hydrogen [({[(2R,3S,4R,5R)-5-(6-amino-9H-purin-9-yl)-3,4-dihydroxyoxolan-2-yl]methoxy}(hydroxy)phosphoryl phosphonato)oxy]phosphonate SMILES: O.O.O.[Na+].[Na+].NC1=C2N=CN([C@@H]3O[C@H](COP(O)(=O)OP([O-])(=O)OP(O)([O-])=O)[C@@H](O)[C@H]3O)C2=NC=N1
Adenosine-5'-triphosphate disodium salt tryhydrate, ≥96%, MP Biomedicals™
CAS: 51963-61-2 Molecular Formula: C10H20N5Na2O16P3 Molecular Weight (g/mol): 605.19 MDL Number: MFCD00150754 InChI Key: MWEQTWJABOLLOS-MVHBFMHHNA-L Synonym: adenosine 5'-triphosphate disodium salt trihydrate,adenosine 5'-triphosphate disodium salt,adenosine 5'-triphosphate disodium salt trihydrate, disodium salt PubChem CID: 134129625 IUPAC Name: disodium trihydrate hydrogen [({[(2R,3S,4R,5R)-5-(6-amino-9H-purin-9-yl)-3,4-dihydroxyoxolan-2-yl]methoxy}(hydroxy)phosphoryl phosphonato)oxy]phosphonate SMILES: O.O.O.[Na+].[Na+].NC1=C2N=CN([C@@H]3O[C@H](COP(O)(=O)OP([O-])(=O)OP(O)([O-])=O)[C@@H](O)[C@H]3O)C2=NC=N1
Sigma Aldrich Adenosine 5'-triphosphate disodium salt trihydrate
MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.