accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 5198-80-1

IUPAC Name: 4-bromo-1,3-thiazole-2-carbaldehyde
Synonyms: 4-bromo-1,3-thiazole-2-carbaldehyde
Molecular Weight (g/mol): 192.03
Molecular Formula: C4H2BrNOS
InChi Key: JDUXMFGFGCJNGO-UHFFFAOYSA-N
SMILES: BrC1=CSC(C=O)=N1
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results