accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3CH3H3COOHNOOCH3

CAS RN 51987-73-6

IUPAC Name: methyl 2-{[(tert-butoxy)carbonyl]amino}-3-phenylpropanoate
Synonyms: methyl 2-[(tert-butoxycarbonyl)amino]-3-phenylpropanoate
Molecular Weight (g/mol): 279.34
Molecular Formula: C15H21NO4
InChi Key: SDSWSVBXRBXPRL-UHFFFAOYNA-N
SMILES: COC(=O)C(CC1=CC=CC=C1)NC(=O)OC(C)(C)C
Molecular Weight (g/mol) 
  • (1)
  • (5)
Quantity 
  • (1)
  • (3)
  • (2)
Percent Purity 
  • (2)
  • (3)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (1)
  • (2)
  • (3)
  • (1)
  • (5)
  • (1)
  • (3)
  • (2)
  • (2)
  • (3)
  • (2)