CAS RN 52-51-7
2-Bromo-2-nitropropane-1,3-diol, 98+%
CAS: 52-51-7 Molecular Formula: C3H6BrNO4 Molecular Weight (g/mol): 199.988 MDL Number: MFCD00007390 InChI Key: LVDKZNITIUWNER-UHFFFAOYSA-N Synonym: bronopol,2-bromo-2-nitro-1,3-propanediol,bronosol,bronocot,bronidiol,bronopolu,bronotak,1,3-propanediol, 2-bromo-2-nitro,onyxide 500,lexgard bronopol PubChem CID: 2450 ChEBI: CHEBI:31306 IUPAC Name: 2-bromo-2-nitropropane-1,3-diol SMILES: C(C(CO)([N+](=O)[O-])Br)O
2-Bromo-2-nitro-1,3-propanediol, MP Biomedicals
CAS: 52-51-7 Molecular Formula: C3H6BrNO4 Molecular Weight (g/mol): 199.988 InChI Key: LVDKZNITIUWNER-UHFFFAOYSA-N Synonym: bronopol,2-bromo-2-nitro-1,3-propanediol,bronosol,bronocot,bronidiol,bronopolu,bronotak,1,3-propanediol, 2-bromo-2-nitro,onyxide 500,lexgard bronopol PubChem CID: 2450 ChEBI: CHEBI:31306 IUPAC Name: 2-bromo-2-nitropropane-1,3-diol SMILES: C(C(CO)([N+](=O)[O-])Br)O
2-Bromo-2-nitro-1,3-propanediol, 98%
CAS: 52-51-7 Molecular Formula: C3H6BrNO4 Molecular Weight (g/mol): 199.99 MDL Number: MFCD00007390 InChI Key: LVDKZNITIUWNER-UHFFFAOYSA-N Synonym: bronopol,2-bromo-2-nitro-1,3-propanediol,bronosol,bronocot,bronidiol,bronopolu,bronotak,1,3-propanediol, 2-bromo-2-nitro,onyxide 500,lexgard bronopol PubChem CID: 2450 ChEBI: CHEBI:31306 IUPAC Name: 2-bromo-2-nitropropane-1,3-diol SMILES: C(C(CO)([N+](=O)[O-])Br)O
2-Bromo-2-nitro-1,3-propanediol, 98.0-100.5%, Spectrum™ Chemical
CAS: 52-51-7 Molecular Formula: C3H6BrNO4 Molecular Weight (g/mol): 199.99 InChI Key: LVDKZNITIUWNER-UHFFFAOYSA-N IUPAC Name: 2-bromo-2-nitropropane-1,3-diol SMILES: OCC(Br)(CO)[N+]([O-])=O
2-Bromo-2-nitro-1,3-propanediol 98.0+%, TCI America™
CAS: 52-51-7 Molecular Formula: C3H6BrNO4 Molecular Weight (g/mol): 199.988 MDL Number: MFCD00007390 InChI Key: LVDKZNITIUWNER-UHFFFAOYSA-N Synonym: bronopol,2-bromo-2-nitro-1,3-propanediol,bronosol,bronocot,bronidiol,bronopolu,bronotak,1,3-propanediol, 2-bromo-2-nitro,onyxide 500,lexgard bronopol PubChem CID: 2450 ChEBI: CHEBI:31306 IUPAC Name: 2-bromo-2-nitropropane-1,3-diol SMILES: C(C(CO)([N+](=O)[O-])Br)O
Sigma Aldrich 2-Bromo-2-nitro-1,3-propanediol
MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.