accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3OPSOCH3OSOONCH3H3C

CAS RN 52-85-7

IUPAC Name: O-4-(dimethylsulfamoyl)phenyl O,O-dimethyl phosphorothioate
Synonyms: famphur warbex rcra waste number P097 fanfos famphur (substance) caswell no. 456D varbex famfur famofos famophos
Molecular Weight (g/mol): 325.33
Molecular Formula: C10H16NO5PS2
InChi Key: JISACBWYRJHSMG-UHFFFAOYSA-N
SMILES: COP(=S)(OC)OC1=CC=C(C=C1)S(=O)(=O)N(C)C

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results