accessibility menu, dialog, popup

UserName

Viewing profile as user:
HONOFCl

CAS RN 52-86-8

IUPAC Name: 4-[4-(4-chlorophenyl)-4-hydroxypiperidin-1-yl]-1-(4-fluorophenyl)butan-1-one
Synonyms: haloperidol novo-peridol mixidol halopal einalon S haldol decanoate keselan bioperidolo halosten aloperidol
Molecular Weight (g/mol): 375.87
Molecular Formula: C21H23ClFNO2
InChi Key: LNEPOXFFQSENCJ-UHFFFAOYSA-N
SMILES: OC1(CCN(CCCC(=O)C2=CC=C(F)C=C2)CC1)C1=CC=C(Cl)C=C1
Molecular Weight (g/mol) 
  • (5)
  • (7)
Percent Purity 
  • (2)
  • (5)
  • (4)
Grade 
  • (1)
  • (9)
Quantity 
  • (1)
  • (1)
  • (2)
  • (3)
Physical Form 
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (4)
  • (5)
  • (1)
  • (4)
  • (2)
  • (1)
  • (5)
  • (7)
  • (2)
  • (5)
  • (4)
  • (1)
  • (9)
  • (1)
  • (1)
  • (2)
  • (3)
  • (2)
  • (1)