accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3OOHOOHO(S)O(R)O(R)O(S)CH3(R)OH(R)OH(R)HO(S)OH(S)HO(R)HOO

CAS RN 520-27-4

IUPAC Name: 5-hydroxy-2-(3-hydroxy-4-methoxyphenyl)-7-{[(2S,3R,4S,5S,6R)-3,4,5-trihydroxy-6-({[(2R,3R,4R,5R,6S)-3,4,5-trihydroxy-6-methyloxan-2-yl]oxy}methyl)oxan-2-yl]oxy}-4H-chromen-4-one
Synonyms: diosmin buchu resin diosmin preparation (product) venosmine 地奥司明 diosmetin 7-O-rutinoside diosmin (substance) diosminum diosmine barosmin
Molecular Weight (g/mol): 608.55
Molecular Formula: C28H32O15
InChi Key: GZSOSUNBTXMUFQ-YFAPSIMESA-N
SMILES: COC1=CC=C(C=C1O)C1=CC(=O)C2=C(O)C=C(O[C@@H]3O[C@H](CO[C@@H]4O[C@@H](C)[C@H](O)[C@@H](O)[C@H]4O)[C@@H](O)[C@H](O)[C@H]3O)C=C2O1
Percent Purity 
  • (2)
  • (2)
Physical Form 
  • (2)
  • (2)
Quantity 
  • (2)
  • (2)
Grade 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)