accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 52009-14-0

IUPAC Name: calcium 2-oxopropanoate
Synonyms: calcium pyruvate
Molecular Weight (g/mol): 127.13
Molecular Formula: C3H3CaO3
InChi Key: QJYMFPMPYYGIJD-UHFFFAOYSA-M
SMILES: [Ca++].CC(=O)C([O-])=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results