accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3COOHOClOCH3

CAS RN 52009-53-7

IUPAC Name: 2-chloro-3,4-dimethoxybenzoic acid
Synonyms: 2-chloro-3,4-dimethoxybenzoic acid
Molecular Weight (g/mol): 216.62
Molecular Formula: C9H9ClO4
InChi Key: YSEIYOHXERRMNN-UHFFFAOYSA-N
SMILES: COC1=C(OC)C(Cl)=C(C=C1)C(O)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results