CAS RN 52009-64-0
IUPAC Name:
6-phenylphenanthridine-3,8-diamine
Synonyms:
6-phenylphenanthridine-3,8-diamine
Molecular Weight (g/mol):
285.35
Molecular Formula:
C19H15N3
InChi Key:
CPNAVTYCORRLMH-UHFFFAOYSA-N
SMILES:
NC1=CC=C2C(=C1)N=C(C1=CC=CC=C1)C1=CC(N)=CC=C21
1
–
1
of
1
results
1
Promotions Available
3,8-Diamino-6-phenylphenanthridine, 98%
3,8-Diamino-6-phenylphenanthridine, 98%
CAS: 52009-64-0 Molecular Formula: C19H15N3 Molecular Weight (g/mol): 285.35 MDL Number: MFCD00075152 InChI Key: CPNAVTYCORRLMH-UHFFFAOYSA-N Synonym: 3,8-diamino-6-phenylphenanthridine,6-phenylphenathridine-3,8-diamine,3,8-dapp,3,8-diamino-6-phenyl-phenanthridine,acmc-1aycj,6-phenyl-3,8-phenanthridinediamine,3,8-diamino-6-phenyl phenanthridine,3,8-phenanthridinediamine,6-phenyl,3,8-diamino-6-phenylphenanthridine6-phenyl-3,8-phenanthridinediamine, PubChem CID: 104052 IUPAC Name: 6-phenylphenanthridine-3,8-diamine SMILES: C1=CC=C(C=C1)C2=C3C=C(C=CC3=C4C=CC(=CC4=N2)N)N