accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CNCH3HNOONHNCH3H3CClClClCl

CAS RN 52022-31-8

IUPAC Name: [(2-chlorophenyl)methyl](2-{[(2-{[(2-chlorophenyl)methyl]diethylazaniumyl}ethyl)carbamoyl]formamido}ethyl)diethylazanium dichloride
Synonyms: ambenonium chloride cloruro de ambenonio chlorure d'ambenonium ambenonii chloridum ambenonium dichloride 安贝氯铵 ambestigmin chloride
Molecular Weight (g/mol): 608.47
Molecular Formula: C28H42Cl4N4O2
InChi Key: DXUUXWKFVDVHIK-UHFFFAOYSA-N
SMILES: [Cl-].[Cl-].CC[N+](CC)(CCNC(=O)C(=O)NCC[N+](CC)(CC)CC1=CC=CC=C1Cl)CC1=CC=CC=C1Cl

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results