accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 52034-92-1

OHO

CAS RN 52034-92-1

IUPAC Name: 2,2-dicyclohexylacetic acid
Synonyms: dicyclohexylacetic acid
Molecular Weight (g/mol): 224.34
Molecular Formula: C14H24O2
InChi Key: PGGMEZOUAPIYOY-UHFFFAOYSA-N
SMILES: OC(=O)C(C1CCCCC1)C1CCCCC1
Molecular Weight (g/mol) 
  • (1)
  • (2)
Percent Purity 
  • (2)
  • (1)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results