accessibility menu, dialog, popup

UserName

Viewing profile as user:
HOOBr

CAS RN 52062-92-7

IUPAC Name: 4-(2-bromoethyl)benzoic acid
Synonyms: 4-(2-bromoethyl)benzoic acid
Molecular Weight (g/mol): 229.07
Molecular Formula: C9H9BrO2
InChi Key: BKMRWJWLBHHGCF-UHFFFAOYSA-N
SMILES: OC(=O)C1=CC=C(CCBr)C=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results