CAS RN 52068-30-1
IUPAC Name:
hydrogen [4-(benzyloxy)phenyl]hydrazine chloride
Synonyms:
hydrogen [4-(benzyloxy)phenyl]hydrazine chloride
Molecular Weight (g/mol):
250.73
Molecular Formula:
C13H15ClN2O
InChi Key:
OVNUPJXMCMTQCN-UHFFFAOYSA-N
SMILES:
[H+].[Cl-].NNC1=CC=C(OCC2=CC=CC=C2)C=C1
1
–
1
of
1
results
1
Promotions Available
4-Benzyloxyphenylhydrazine hydrochloride, 98%, Thermo Scientific Chemicals
4-Benzyloxyphenylhydrazine hydrochloride, 98%, Thermo Scientific Chemicals
CAS: 52068-30-1 Molecular Formula: C13H14N2O·HCl Molecular Weight (g/mol): 250.73 MDL Number: MFCD00137741 InChI Key: OVNUPJXMCMTQCN-UHFFFAOYSA-N Synonym: 4-benzyloxyphenylhydrazine hydrochloride,4-benzyloxy phenyl hydrazine hydrochloride,4-benzyloxyphenyl hydrazine hydrochloride,4-benzyloxy phenyl hydrazine monohydrochloride,4-benzyloxyphenyl hydrazine hcl,4-benzyloxyphenylhydrazine hcl,4-benzyloxyphenylhydrazinehydrochloride,4-phenylmethoxyphenyl hydrazine hydrochloride PubChem CID: 3016725 IUPAC Name: (4-phenylmethoxyphenyl)hydrazine;hydrochloride SMILES: C1=CC=C(C=C1)COC2=CC=C(C=C2)NN.Cl