accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 52070-67-4

IUPAC Name: 1-(prop-2-yn-1-yl)piperazine
Synonyms: 1-(prop-2-yn-1-yl)piperazine
Molecular Weight (g/mol): 124.19
Molecular Formula: C7H12N2
InChi Key: GWCSATTUAOHJDK-UHFFFAOYSA-N
SMILES: C#CCN1CCNCC1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results