CAS RN 52092-43-0
IUPAC Name:
2-bromo-4-nitropyridin-1-ium-1-olate
Synonyms:
2-bromo-4-nitropyridin-1-ium-1-olate
Molecular Weight (g/mol):
218.99
Molecular Formula:
C5H3BrN2O3
InChi Key:
IRBDHXCXCSFNEQ-UHFFFAOYSA-N
SMILES:
[O-][N+](=O)C1=CC=[N+]([O-])C(Br)=C1
1
–
1
of
1
results
1
2-Bromo-4-nitropyridine N-Oxide 98.0+%, TCI America™
CAS: 52092-43-0 Molecular Formula: C5H3BrN2O3 Molecular Weight (g/mol): 218.99 MDL Number: MFCD00160743 InChI Key: IRBDHXCXCSFNEQ-UHFFFAOYSA-N PubChem CID: 3904026 IUPAC Name: 2-bromo-4-nitropyridin-1-ium-1-olate SMILES: [O-][N+](=O)C1=CC=[N+]([O-])C(Br)=C1