accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3OH3CO

CAS RN 5211-62-1

IUPAC Name: 1-(2-methoxyphenyl)propan-2-one
Synonyms: 1-(2-methoxyphenyl)propan-2-one
Molecular Weight (g/mol): 164.2
Molecular Formula: C10H12O2
InChi Key: GMBFNZCPZFVKAT-UHFFFAOYSA-N
SMILES: COC1=CC=CC=C1CC(C)=O
Boiling Point 
  • (2)
  • (1)
Percent Purity 
  • (2)
  • (1)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (2)
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)