accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 52126-51-9

IUPAC Name: sodium 5-oxo-1-(4-sulfophenyl)-4,5-dihydro-1H-pyrazole-3-carboxylate
Synonyms: sodium 5-oxo-1-(4-sulfophenyl)-4H-pyrazole-3-carboxylate
Molecular Weight (g/mol): 306.22
Molecular Formula: C10H7N2NaO6S
InChi Key: KFVXGVBBXSOYNO-UHFFFAOYSA-M
SMILES: [Na+].OS(=O)(=O)C1=CC=C(C=C1)N1N=C(CC1=O)C([O-])=O
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results