accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 52152-93-9

IUPAC Name: sodium 4-carbamoyl-1-{[(6R,7R)-2-carboxylato-8-oxo-7-[(2R)-2-phenyl-2-sulfonatoacetamido]-5-thia-1-azabicyclo[4.2.0]oct-2-en-3-yl]methyl}pyridin-1-ium
Synonyms: cefsulodin
Molecular Weight (g/mol): 554.52
Molecular Formula: C22H19N4NaO8S2
InChi Key: REACMANCWHKJSM-DWBVFMGKSA-M
SMILES: [Na+].NC(=O)C1=CC=[N+](CC2=C(N3[C@H](SC2)[C@H](NC(=O)[C@@H](C2=CC=CC=C2)S([O-])(=O)=O)C3=O)C([O-])=O)C=C1
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
Physical Form 
  • (1)
  • (2)
Type 
  • (3)
Color 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results