accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 52178-50-4

IUPAC Name: methyl 3-formylbenzoate
Synonyms: methyl 3-formylbenzoate
Molecular Weight (g/mol): 164.16
Molecular Formula: C9H8O3
InChi Key: UVSBCUAQEZINCQ-UHFFFAOYSA-N
SMILES: COC(=O)C1=CC=CC(C=O)=C1
Molecular Weight (g/mol) 
  • (1)
  • (3)
Percent Purity 
  • (1)
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (1)
  • (3)
  • (1)
  • (3)
  • (1)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)