accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3NNNN

CAS RN 52197-12-3

IUPAC Name: 5-methylpyrazine-2,3-dicarbonitrile
Synonyms: 5-methylpyrazine-2,3-dicarbonitrile
Molecular Weight (g/mol): 144.14
Molecular Formula: C7H4N4
InChi Key: RHYUBLSWHDYKAO-UHFFFAOYSA-N
SMILES: CC1=CN=C(C#N)C(=N1)C#N
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results