accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 522-75-8

IUPAC Name: 2-[(2Z)-3-oxo-2,3-dihydro-1-benzothiophen-2-ylidene]-2,3-dihydro-1-benzothiophen-3-one
Synonyms: thioindigo
Molecular Weight (g/mol): 296.36
Molecular Formula: C16H8O2S2
InChi Key: JOUDBUYBGJYFFP-NXVVXOECSA-N
SMILES: O=C1\C(SC2=CC=CC=C12)=C1\SC2=CC=CC=C2C1=O
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results