CAS RN 52204-65-6
IUPAC Name:
4-propylcyclohexan-1-ol
Synonyms:
4-propylcyclohexan-1-ol
Molecular Weight (g/mol):
142.24
Molecular Formula:
C9H18O
InChi Key:
YVPZFPKENDZQEJ-UHFFFAOYSA-N
SMILES:
CCCC1CCC(O)CC1
1
–
1
of
1
results
1
4-Propylcyclohexanol (cis- and trans- mixture) 98.0+%, TCI America™
CAS: 52204-65-6 Molecular Formula: C9H18O Molecular Weight (g/mol): 142.242 MDL Number: MFCD00091618 InChI Key: YVPZFPKENDZQEJ-UHFFFAOYSA-N PubChem CID: 2775094 IUPAC Name: 4-propylcyclohexan-1-ol SMILES: CCCC1CCC(CC1)O