accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 52206-05-0

IUPAC Name: 1-acetyl-2,3-dihydro-1H-indole-5-sulfonyl chloride
Synonyms: 1-acetyl-2,3-dihydroindole-5-sulfonyl chloride
Molecular Weight (g/mol): 259.7
Molecular Formula: C10H10ClNO3S
InChi Key: QNFXLCHANYHGIF-UHFFFAOYSA-N
SMILES: CC(=O)N1CCC2=C1C=CC(=C2)S(Cl)(=O)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results