accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 52267-39-7

IUPAC Name: 1-benzyl 3-methyl propanedioate
Synonyms: 1-benzyl 3-methyl propanedioate
Molecular Weight (g/mol): 208.21
Molecular Formula: C11H12O4
InChi Key: IAUZDBFOEWAQFE-UHFFFAOYSA-N
SMILES: COC(=O)CC(=O)OCC1=CC=CC=C1
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (2)
  • (2)
Boiling Point 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)