accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CNNNOO

CAS RN 5228-48-8

IUPAC Name: 2-methyl-5-nitro-2H-indazole
Synonyms: 2-methyl-5-nitroindazole
Molecular Weight (g/mol): 177.16
Molecular Formula: C8H7N3O2
InChi Key: IWPBWVZEOVCZRX-UHFFFAOYSA-N
SMILES: CN1C=C2C=C(C=CC2=N1)[N+]([O-])=O
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results