accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 523-21-7

IUPAC Name: disodium tetraoxocyclohex-1-ene-1,2-bis(olate)
Synonyms: sodium rhodizonate
Molecular Weight (g/mol): 214.04
Molecular Formula: C6Na2O6
InChi Key: SXWPCWBFJXDMAI-UHFFFAOYSA-L
SMILES: [Na+].[Na+].[O-]C1=C([O-])C(=O)C(=O)C(=O)C1=O
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results