accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 523-42-2

IUPAC Name: 1-(3-methylbutyl)-4-{[1-(3-methylbutyl)-1,4-dihydroquinolin-4-ylidene]methyl}quinolin-1-ium iodide
Synonyms: 1-(3-methylbutyl)-4-{[1-(3-methylbutyl)quinolin-4-ylidene]methyl}quinolin-1-ium iodide
Molecular Weight (g/mol): 538.52
Molecular Formula: C29H35IN2
InChi Key: QGKMIGUHVLGJBR-UHFFFAOYSA-M
SMILES: [I-].CC(C)CCN1C=CC(=CC2=C3C=CC=CC3=[N+](CCC(C)C)C=C2)C2=CC=CC=C12
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results