accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 523-50-2

OOO

CAS RN 523-50-2

IUPAC Name: 2H-furo[2,3-h]chromen-2-one
Synonyms: angelicin furo(5',4':7,8)coumarin angecin isopsoralen angelicin (coumarin deriv) angelicin (coumarin derivative)
Molecular Weight (g/mol): 186.17
Molecular Formula: C11H6O3
InChi Key: XDROKJSWHURZGO-UHFFFAOYSA-N
SMILES: O=C1OC2=C3C=COC3=CC=C2C=C1
Percent Purity 
  • (1)
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results