accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 52359-17-8

IUPAC Name: bis(di-tert-butyl(phenyl)phosphane) palladium
Synonyms: bis(di-tert-butyl(phenyl)phosphane) palladium
Molecular Weight (g/mol): 551.04
Molecular Formula: C28H46P2Pd
InChi Key: KJTQUBRAVOMHKY-UHFFFAOYSA-N
SMILES: [Pd].CC(C)(C)P(C1=CC=CC=C1)C(C)(C)C.CC(C)(C)P(C1=CC=CC=C1)C(C)(C)C
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results