accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 52390-26-8

IUPAC Name: 2-benzoyl-decahydroisoquinolin-6-one
Synonyms: 2-benzoyl-octahydroisoquinolin-6-one
Molecular Weight (g/mol): 257.33
Molecular Formula: C16H19NO2
InChi Key: OSZFDJVZOKTWRW-UHFFFAOYNA-N
SMILES: O=C(N1CCC2CC(=O)CCC2C1)C1=CC=CC=C1
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results