accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 52395-66-1

IUPAC Name: (isothiocyanatomethyl)cyclohexane
Synonyms: (isothiocyanatomethyl)cyclohexane
Molecular Weight (g/mol): 155.26
Molecular Formula: C8H13NS
InChi Key: RYBJNWIQNLZNPE-UHFFFAOYSA-N
SMILES: S=C=NCC1CCCCC1
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results