accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CONOCH3

CAS RN 524-96-9

IUPAC Name: benzyl[bis(4-methoxyphenyl)methylidene]amine
Synonyms: benzyl[bis(4-methoxyphenyl)methylidene]amine
Molecular Weight (g/mol): 331.42
Molecular Formula: C22H21NO2
InChi Key: IZZONQFQTGCWNC-UHFFFAOYSA-N
SMILES: COC1=CC=C(C=C1)C(=NCC1=CC=CC=C1)C1=CC=C(OC)C=C1
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results