accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 5241-58-7

IUPAC Name: (2S)-2-amino-3-phenylpropanamide
Synonyms: phenylalanine amide PheNH2 L-Phe-NH2 Phe-NH2 H-Phe-NH2
Molecular Weight (g/mol): 164.21
Molecular Formula: C9H12N2O
InChi Key: OBSIQMZKFXFYLV-QMMMGPOBSA-N
SMILES: N[C@@H](CC1=CC=CC=C1)C(N)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (1)
  • (1)