accessibility menu, dialog, popup

UserName

Viewing profile as user:
OHNNHNNS

CAS RN 52431-78-4

IUPAC Name: 1-(4-hydroxyphenyl)-2,5-dihydro-1H-1,2,3,4-tetrazole-5-thione
Synonyms: 1-(4-hydroxyphenyl)-2H-1,2,3,4-tetrazole-5-thione
Molecular Weight (g/mol): 194.21
Molecular Formula: C7H6N4OS
InChi Key: MOXZSKYLLSPATM-UHFFFAOYSA-N
SMILES: OC1=CC=C(C=C1)N1NN=NC1=S
Percent Purity 
  • (2)
  • (3)
Quantity 
  • (1)
  • (2)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (2)
  • (3)
  • (2)
  • (3)
  • (1)
  • (2)
  • (2)
  • (2)