accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3N(R)(S)NH2(S)OHOOHNO

CAS RN 52450-18-7

IUPAC Name: 1-[(2R,4S,5S)-4-amino-5-(hydroxymethyl)oxolan-2-yl]-5-methyl-1,2,3,4-tetrahydropyrimidine-2,4-dione
Synonyms: 3'-amino-3'-deoxythymidine
Molecular Weight (g/mol): 241.25
Molecular Formula: C10H15N3O4
InChi Key: ADVCGXWUUOVPPB-XLPZGREQSA-N
SMILES: CC1=CN([C@H]2C[C@H](N)[C@@H](CO)O2)C(=O)NC1=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results