CAS RN 524674-17-7
CAS RN 524674-17-7
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Thermo Scientific Chemicals 2-(2-Pyrrolidinyl)thiazole, 97%, Thermo Scientific™
CAS: 524674-17-7 Molecular Formula: C7H10N2S MDL Number: MFCD04966887 Synonym: 2-(2-Pyrrolidino)thiazole; 2-(2-Thiazolyl)pyrrolidine