CAS RN 524955-09-7
IUPAC Name:
3-chloro-4-[(pyridin-2-yl)methoxy]aniline
Synonyms:
3-chloro-4-(pyridin-2-ylmethoxy)aniline
Molecular Weight (g/mol):
234.68
Molecular Formula:
C12H11ClN2O
InChi Key:
XCAPJQSICQSUJP-UHFFFAOYSA-N
SMILES:
NC1=CC(Cl)=C(OCC2=CC=CC=N2)C=C1
1
–
1
of
1
results
1
3-Chloro-4-(2-pyridylmethoxy)aniline 98.0+%, TCI America™
CAS: 524955-09-7 Molecular Formula: C12H11ClN2O Molecular Weight (g/mol): 234.68 MDL Number: MFCD08699389 InChI Key: XCAPJQSICQSUJP-UHFFFAOYSA-N PubChem CID: 16781212 IUPAC Name: 3-chloro-4-[(pyridin-2-yl)methoxy]aniline SMILES: NC1=CC(Cl)=C(OCC2=CC=CC=N2)C=C1