accessibility menu, dialog, popup

UserName

Viewing profile as user:
FOHOI

CAS RN 52548-63-7

IUPAC Name: 5-fluoro-2-iodobenzoic acid
Synonyms: 5-fluoro-2-iodobenzoic acid
Molecular Weight (g/mol): 266.01
Molecular Formula: C7H4FIO2
InChi Key: XPFMQYOPTHMSJJ-UHFFFAOYSA-N
SMILES: OC(=O)C1=C(I)C=CC(F)=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results