accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 52558-16-4

IUPAC Name: disodium [(2R,3S,5R)-5-(2-amino-6-oxo-6,9-dihydro-1H-purin-9-yl)-3-hydroxyoxolan-2-yl]methyl phosphate
Synonyms: disodium 2'-deoxyguanosine 5'-phosphate
Molecular Weight (g/mol): 391.19
Molecular Formula: C10H12N5Na2O7P
InChi Key: CTPAMSRBXKGZCJ-WJQTVUBHNA-L
SMILES: [Na+].[Na+].NC1=NC2=C(N=CN2[C@H]2C[C@H](O)[C@@H](COP([O-])([O-])=O)O2)C(=O)N1
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results