accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 5257-24-9

IUPAC Name: 3-methyl-1H-indole-2-carbaldehyde
Synonyms: 3-methyl-1H-indole-2-carbaldehyde
Molecular Weight (g/mol): 159.19
Molecular Formula: C10H9NO
InChi Key: GHSUORIVDBPKKQ-UHFFFAOYSA-N
SMILES: CC1=C(NC2=CC=CC=C12)C=O
Molecular Weight (g/mol) 
  • (1)
  • (2)
Percent Purity 
  • (1)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (2)