CAS RN 5259-50-7
IUPAC Name:
methyl (1S,2R,4R)-1-methoxybicyclo[2.2.2]oct-5-ene-2-carboxylate
Synonyms:
methyl (1S,2R,4R)-1-methoxybicyclo[2.2.2]oct-5-ene-2-carboxylate
Molecular Weight (g/mol):
196.25
Molecular Formula:
C11H16O3
InChi Key:
DFEJFZWTBMJTQN-WCABBAIRSA-N
SMILES:
COC(=O)[C@@H]1C[C@H]2CC[C@]1(OC)C=C2
1
–
1
of
1
results
1
Methyl 1-methoxybicyclo[2.2.2]oct-5-ene-2-carboxylate, 95%, mixture of endo and exo, Thermo Scientific™
CAS: 5259-50-7 Molecular Formula: C11H16O3 Molecular Weight (g/mol): 196.246 InChI Key: DFEJFZWTBMJTQN-UHFFFAOYSA-N Synonym: methyl 1-methoxybicyclo 2.2.2 oct-5-ene-2-carboxylate,1-methoxybicyclo 2.2.2 oct-5-ene-2-carboxylic acid methyl ester,acmc-20akjd,methyl 4-methoxybicyclo 2.2.2 oct-2-ene-5-carboxylate,bicyclo 2.2.2 oct-5-ene-2-carboxylicacid, 1-methoxy-, methyl ester PubChem CID: 2724151 IUPAC Name: methyl 4-methoxybicyclo[2.2.2]oct-2-ene-5-carboxylate SMILES: COC(=O)C1CC2CCC1(C=C2)OC