accessibility menu, dialog, popup

UserName

Viewing profile as user:
HOHN

CAS RN 526-55-6

IUPAC Name: 2-(1H-indol-3-yl)ethan-1-ol
Synonyms: tryptophol|dl-tryptophanol|IEA
Molecular Weight (g/mol): 161.2
Molecular Formula: C10H11NO
InChi Key: MBBOMCVGYCRMEA-UHFFFAOYSA-N
SMILES: OCCC1=CNC2=CC=CC=C12
Molecular Weight (g/mol) 
  • (1)
  • (7)
Percent Purity 
  • (3)
  • (5)
Quantity 
  • (3)
  • (1)
  • (3)
  • (1)
Boiling Point 
  • (2)
  • (2)
  • (3)
Physical Form 
  • (2)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (3)
  • (4)
  • (1)
  • (7)
  • (3)
  • (5)
  • (3)
  • (1)
  • (3)
  • (1)
  • (2)
  • (2)
  • (3)
  • (2)
  • (3)