accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 526-73-8

IUPAC Name: 1,2,3-trimethylbenzene
Synonyms: 1,2,3-trimethylbenzene hemimellitene hemimellitol hemellitol
Molecular Weight (g/mol): 120.2
Molecular Formula: C9H12
InChi Key: FYGHSUNMUKGBRK-UHFFFAOYSA-N
SMILES: CC1=CC=CC(C)=C1C
Molecular Weight (g/mol) 
  • (2)
  • (3)
Boiling Point 
  • (2)
  • (2)
Quantity 
  • (1)
  • (2)
  • (1)
Percent Purity 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (2)
  • (2)
  • (3)
  • (2)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (2)
  • (1)