accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3HOH3C

CAS RN 526-75-0

IUPAC Name: 2,3-dimethylphenol
Synonyms: 2,3-dimethylphenol
Molecular Weight (g/mol): 122.17
Molecular Formula: C8H10O
InChi Key: QWBBPBRQALCEIZ-UHFFFAOYSA-N
SMILES: CC1=CC=CC(O)=C1C
Molecular Weight (g/mol) 
  • (2)
  • (3)
Boiling Point 
  • (2)
  • (2)
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (1)
  • (1)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (2)
  • (2)
  • (3)
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
  • (2)
  • (2)
  • (1)